Learn More
Abnova™ Human MAP4 Full-length ORF (NP_112147.2, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004134-P01.25ug
Description
The protein encoded by this gene is a major non-neuronal microtubule-associated protein. This protein contains a domain similar to the microtubule-binding domains of neuronal microtubule-associated protein (MAP2) and microtubule-associated protein tau (MAPT/TAU). This protein promotes microtubule assembly, and has been shown to counteract destabilization of interphase microtubule catastrophe promotion. Cyclin B was found to interact with this protein, which targets cell division cycle 2 (CDC2) kinase to microtubules. The phosphorylation of this protein affects microtubule properties and cell cycle progression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MADLSLADALTEPSPDIEGEIKRDFIATLEAEAFDDVVGETVGKTDYIPLLDVDEKTGNSESKKKPCSETSQIEDTPSSKPTLLANGGHGVEGSDTTEASpecifications
NP_112147.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779A1753/MGC8617 | |
MAP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4134 | |
MAP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADLSLADALTEPSPDIEGEIKRDFIATLEAEAFDDVVGETVGKTDYIPLLDVDEKTGNSESKKKPCSETSQIEDTPSSKPTLLANGGHGVEGSDTTEA | |
RUO | |
MAP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |