missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Plexin B2 Protein, His Tag

Specifications

Molecular Weight (g/mol) 126.9 kDa
Source Human Plexin B2, His Tag (PL2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 1160 (Accession # O15031-1).
Immunogen Leu 20 - Arg 1160
Storage Requirements -20°C
Gene Alias Plexin B2,PLXNB2,KIAA0315,MM1,Plexin-B2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17206420
Encompass_Preferred
View Documents Promotion Details
ACROBiosystems
PL2-H52H3-100ug
100 μg N/A
View alternative products
Save Up to 
»
N/A
17216420
Encompass_Preferred
View Documents Promotion Details
ACROBiosystems
PL2-H52H3-1mg
1 mg N/A
View alternative products
Save Up to 
»
N/A
Description

Description

Plexin B2 serves as the receptor of Sema4C and Sema4G. By signaling the effect of Sema4C and Sema4G, the plexin B2 receptor plays important roles in neural tube closure and cerebellar granule cell development. Mice lacking Plexin B2 demonstrated defects in closure of the neural tube and disorganization of the embryonic brain. In developing kidney, Sema4C-Plexin B2 signaling modulates ureteric branching. Plexin B2 is expressed both in the pretubular aggregates and the ureteric epithelium in the developing kidney. Deletion of Plexin B2 results in renal hypoplasia and occasional double ureters. The Sema domain is located at the N-terminus and contains four disulfide bonds formed by eight conserved cysteine residues. It serves as a ligand-recognition and -binding module.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 126.9 kDa.
Specifications

Specifications

126.9 kDa
Leu 20 - Arg 1160
Plexin B2,PLXNB2,KIAA0315,MM1,Plexin-B2
Unconjugated
His Tag
Powder
Human Plexin B2, His Tag (PL2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 1160 (Accession # O15031-1).
-20°C
Plexin B2
Human
HEK293
Other Recombinant Proteins
Videos
SDS
Documents

Documents

Product Certifications