missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ST18 Partial ORF (NP_055497, 201 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009705-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: NGWDSGSNFSEETKPPRVPKYVLTDHKKDLLEVPEIKTEGDKFIPCENRCDSETERKDPQNALAEPLDGNAQPSFPDVEEEDSESLAVMTEEGSDLEKAKSpecifications
NP_055497 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NGWDSGSNFSEETKPPRVPKYVLTDHKKDLLEVPEIKTEGDKFIPCENRCDSETERKDPQNALAEPLDGNAQPSFPDVEEEDSESLAVMTEEGSDLEKAK | |
RUO | |
ST18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9705 | |
ST18 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0535/ZNF387 | |
ST18 | |
Recombinant | |
wheat germ expression system |