missing translation for 'onlineSavingsMsg'
Learn More
Ansell HyFlex™ 11-917 Partial Nitrile-Coated Seamless Knit Liquid Repellant Gloves Product Code.: 16059681
Estimated Shipment: 31-07-2025
Only null left
Add to basket

Ansell HyFlex™ 11-917 Partial Nitrile-Coated Seamless Knit Liquid Repellant Gloves
GREENER_CHOICE

Product Code. 16059681 Shop All Ansell Products
9, 12 pairs
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16059681

Brand: Ansell 11917090

Only null left
Estimated Shipment: 31-07-2025
Add to basket
Only null left
Add to basket
This item is not returnable. View return policy

Gloves repel oil, grease, and dirt making them ideal for use in grimey work environments. Seamless nylon liner provides comfort, dexterity, and a secure fit.

  • Liquid repellent against oil, grease, and dirt
  • Seamless nylon liner
  • Suitable for food handling
Greener Choice Claims
  • Environmental benefits include:
    • Free-of a substance that causes environmental harm
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Exterior Finish Rough
Hand Specific
Latex Free No
Material Nylon
Product Type Mechanical Protection Gloves
Size 9
EN388 Standard 4121A
For Use With (Application) Maintenance, Quality Control, and Metal Fabrication
Glove Cuff Color White
Glove Cuff Style Knit
Series 11-917
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell HyFlex™ 11-917 Partial Nitrile-Coated Seamless Knit Liquid Repellant Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.