Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Collection Rack
Select a 16- or 24-position collection rack for your Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold.
Brand: Thermo Scientific™ 60104-255
83.61 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Select a 16- or 24-position collection rack for use with the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold. The manifold enables solid phase extraction sample preparation using SPE cartridges. Extracts are collected into tubes in the collection rack.
HyperSep Glass Block Manifolds are durable, chemically resistant units designed to provide years of trouble-free extractions.Polypropylene racks are highly resistant to chemical degradation and abuse, and are supplied according to the number of positions on the manifold lid
Vacuum control valve allows optimization of sample flow rates during extraction
Test tube racks allow the use of 12-13mm or 16mm disposable test tubes
Vacuum gauge and bleed valve enables easy monitoring of the vacuum process
Corner leg supports prevent damage to the luers and tips when the lid is not being used
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
Retaining Clips for Collection Rack, 12 pk | |
Collection Rack for Glass Block Vacuum Manifold | |
Retaining Clips |
Extract Collection | |
HyperSep |