Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Collection Rack
Select a 16- or 24-position collection rack for your Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold.
117.00€ - 462.00€
Specifications
For Use With (Application) Extract Collection
Product Line HyperSep
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10361245
View Documents
Thermo Scientific™
60104-251
Collection Rack for 16 Port Vacuum Manifold, 1 pk
462.00€
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
10289454
View Documents
Thermo Scientific™
60104-252
Collection Rack for 24 Port Vacuum Manifold, 1 pk This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
10611226
View Documents
Thermo Scientific™
60104-255
Retaining Clips for Collection Rack, 12 pk
117.00€
Pack of 12
Only null left
Estimated Shipment: 15-07-2025
Add to basket
Description
Description
Select a 16- or 24-position collection rack for use with the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold. The manifold enables solid phase extraction sample preparation using SPE cartridges. Extracts are collected into tubes in the collection rack.
HyperSep Glass Block Manifolds are durable, chemically resistant units designed to provide years of trouble-free extractions.
Polypropylene racks are highly resistant to chemical degradation and abuse, and are supplied according to the number of positions on the manifold lid
Vacuum control valve allows optimization of sample flow rates during extraction
Test tube racks allow the use of 12-13mm or 16mm disposable test tubes
Vacuum gauge and bleed valve enables easy monitoring of the vacuum process
Corner leg supports prevent damage to the luers and tips when the lid is not being used
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
Specifications
Extract Collection
HyperSep
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Collection Rack