Learn More
DiscoverX InCELL Hunter™ HEK 293 BRD3(1,2) Bromodomain Cell Line
The InCELL Hunter™ HEK 293 BRD3(1,2) Bromodomain Cell Line measures target engagement via compound-induced stabilization of BRD3(1,2) target protein. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 96-0038C1
This item is not returnable.
View return policy
Description
The InCELL Hunter™ HEK 293 BRD3(1,2) Bromodomain Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of BRD3(1,2) via detection of BRD3(1,2) levels. This product uses EFC technology. The cell line included overexpresses BRD3(1,2) target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Target Engagement | |
Cell Lines | |
2 Vials | |
NM_007371.3 |
293H | |
(+)-JQ1 | |
BRD3(1,2) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.