missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD 20nm Midi Antibody Labeling Kit

Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.

Brand:  Novus Biologicals™ 229-0015

Product Code. 18010196

  • 992.00€ / 1 test
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

For Research Use Only

This item is not returnable. View return policy

Description

Description

Features
  • Proprietary surface coat reduces risk of sub-optimal antibody performance
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Antibody-GOLD conjugates in just 15 minutes
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Coating procedure not dependent on isoelectric point of antibody
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services
Specifications

Specifications

Store at 4°C. Do not freeze.
1 Test
Antibody Labeling Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers