Learn More
Cole-Parmer™ Jumbo Holder
Jumbo Holder is ideal for holding clamps or ring stands to lab-frames at a 90° angle. Connectors and holders do not include rods (Order separately).
Brand:  Cole-Parmer™ 08041-22
Product Code. 11890020
  • 49.65€ / Each
Estimated Shipment: 23-05-2025
Description
Jumbo Holder is ideal for holding clamps or ring stands to lab-frames at a 90° angle. Connectors and holders do not include rods (Order separately).
Specifications
Specifications
1 Ea.
Aluminum
1/2 in. (12.7 mm) Diameter Frame Rods
Jumbo Holder
0 to 0.83 in. diameter rods
Product Title
Cole-Parmer™ Jumbo Holder > jumbo holder