Learn More
Kartell™ Plastilab™ Two-Tier Holder for Vertical Pipette Stand
Consists of two-tiered pipette holder only, base and support rod are not included.
Brand: Kartell™ 0026500
31.79 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Consists of two-tiered pipette holder only, base and support rod are not included.
- Made from polypropylene
- Boss head allows holder to be raised or lowered on support rod
Specifications
Pipet Holder | |
18 Small and 10 Large Holes Places | |
175 mm |
Polypropylene | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 13
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
27Hours
18Minutes
42Seconds
4
Product Content Correction
Product Title
Kartell™ Plastilab™ Two-Tier Holder for Vertical Pipette Stand > Holds: 18 Small and 10 Large Holes Places