Learn More
DiscoverX KILR™ CD16 Effector Cells
The KILR™ CD16 Effector Cells are used with KILR cell lines and pools to induce assayable cell death. These effector cells ship as a single vial of cryopreserved primary cells.
Brand: DiscoverX 97-0007-01
This item is not returnable.
View return policy
Description
This product is a companion product for use with KILR cell lines and pools. These KILR CD16 Effector Cells are purified human cytotoxic T lymphocytes stably expressing CD16, intended for use as effector cells in antibody-dependent cell-mediated cytotoxicity (ADCC) assays assessing cell death. The cells are produced in a standardized batch to allow for high levels of assay reproducibility. These purified effector cells are provided as one vial of cryopreserved cells.Specifications
Cytotoxic | |
Effector Cell | |
1 Vial |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.