Learn More
DiscoverX KILR™ CD16 Effector Cells
The KILR™ CD16 Effector Cells are used with KILR cell lines and pools to induce assayable cell death. These effector cells ship as a single vial of cryopreserved primary cells.
Brand:  DiscoverX 97-0007-01
Encompass
Product Code. 17324839
  • 2248.50€ / 1 ampoule
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This product is a companion product for use with KILR cell lines and pools. These KILR CD16 Effector Cells are purified human cytotoxic T lymphocytes stably expressing CD16, intended for use as effector cells in antibody-dependent cell-mediated cytotoxicity (ADCC) assays assessing cell death. The cells are produced in a standardized batch to allow for high levels of assay reproducibility. These purified effector cells are provided as one vial of cryopreserved cells.
Specifications
Specifications
Cytotoxic
Effector Cell
1 Vial