missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Free Base, Fisher BioReagents

65.50€ - 298.00€

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10041693
View Documents
Fisher BioReagents
BP2505-100
100 g Poly Bottle This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
10421725
View Documents
Fisher BioReagents
BP2505-500
500 g Poly Bottle This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
10368364
View Documents
Fisher BioReagents
BP2505-1
1 kg Poly Bottle This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

L-Arginine is an essential amino acid used in biochemical research applications.
Specifications

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
6322
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
C6H14N4O2
MFCD00002635
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
CHEBI:16467
NC(CCCN=C(N)N)C(O)=O

Specifications

223°C
11.4
≥99 %
C6H14N4O2
MFCD00002635
15, 770
ODKSFYDXXFIFQN-UHFFFAOYNA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
174.20
CHEBI:16467
≥99%
L-Arginine, Free Base
74-79-3
White
Pass Test
Conforms to reference
0.3% max.
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
NC(CCCN=C(N)N)C(O)=O
+26.3 to +27.7° (+ or -)
6322
174.12
Solid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

Product Identifier
  • L(+)-Arginine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.