missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford L-Arginine HCl
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford A50400-500.0
This item is not returnable.
View return policy
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 1119-34-2
- Molecular Formula: C6H14N4O2 HCl
- Molecular Weight: 210.7
Specifications
L-Arginine HCl | |
1119-34-2 | |
C6H14N4O2-HCl | |
500 g |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Melford L-Arginine HCl > 500 g
Spot an opportunity for improvement?Share a Content Correction