Learn More
Melford L-Arginine HCl
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
58.15€ - 1660.00€
Specifications
Chemical Name or Material | L-Arginine HCl |
---|---|
CAS | 1119-34-2 |
Molecular Formula | C6H14N4O2-HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018045
|
Melford
A50400-100.0 |
100 g |
58.15€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018048
|
Melford
A50400-5000.0 |
5 kg |
1660.00€
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018046
|
Melford
A50400-1000.0 |
1 kg |
407.00€
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018047
|
Melford
A50400-500.0 |
500 g |
210.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 1119-34-2
- Molecular Formula: C6H14N4O2 HCl
- Molecular Weight: 210.7
Specifications
L-Arginine HCl | |
1119-34-2 | |
C6H14N4O2-HCl |
Product Title
Melford L-Arginine HCl