Learn More
OHAUS™ Large Two Prong Sleeves
OHAUS™ Large Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand: OHAUS™ 30400255
9.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in red vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
Specifications
2 | |
Vinyl | |
Red |
Prong Sleeve | |
Large |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
OHAUS™ Large Two Prong Sleeves > Material: Vinyl