missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Antibody Labeling Kit
Applications: ELISA
481.00€ - 988.00€
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | Alkaline Phosphatase |
Product Type | Alkaline Phosphatase Antibody Labeling Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18038494
|
Novus Biologicals™
702-0005 |
100 ÎĽg |
481.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700404
|
Novus Biologicals™
702-0010 |
300 ÎĽg |
733.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18738793
|
Novus Biologicals™
702-0015 |
1 mg |
988.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18716733
|
Novus Biologicals™
702-0030 |
30 ÎĽg |
535.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alkaline phosphatase (AP) is a hydrolyase enzyme that is frequently conjugated to antibodies for use in immunoassays. PNPP is a popular colorimetric substrate for AP; the product of the reaction, p-nitrophenol, can be read at 405nM. Fluorogenic substrates (e.g. 4-methylumbelliferyl phosphate; MUP) may also be employed.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Alkaline Phosphatase | |
Alkaline Phosphatase Antibody Labeling Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Antibody Labeling Kit