missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit

Applications: Flow Cytometry

Brand:  Novus Biologicals™ 703-0004

 View more versions of this product

Product Code. 18749963

  • 3050.00€ / 3mg
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from red algae. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield.

Order Info

Quantity

Specifications

Specifications

Store at -20°C. Avoid freeze-thaw cycles.
3 mg
PE
R-PE Antibody Labeling Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers