missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid AMCA Antibody Labeling Kit
Antibody labeling kit
544.00€ - 1115.00€
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | AMCA |
Kit Contents | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |
Product Type | Rapid AMCA Antibody Labeling Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18034894
|
Novus Biologicals™
313-0030 |
30 μg |
544.00€
1 set |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
18078284
|
Novus Biologicals™
313-0010 |
300 μg |
761.00€
1 set |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
18034014
|
Novus Biologicals™
313-0015 |
1 mg |
1115.00€
1mg |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
Description
Unlike standard AMCA labeling procedures, where a large molar excess of AMCA is employed, the Lightning-Link AMCA kit combines a low molar ratio of AMCA:antibody and high labeling efficiency to eliminate the need for purification steps. AMCA (aminomethylcoumarin acetate) is a blue fluorescent dye commonly used in fluorescence microscopy, arrays and in-situ hybridisation. It has an excitation wavelength of 345-350nm and an emission wavelength of 440-460nm.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
AMCA | |
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent | |
Rapid AMCA Antibody Labeling Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title