missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Rhodamine Antibody Labeling Kit

Label antibodies, proteins, peptides and other biomolecules with rhodamine, using simple procedure. Set up conjugations in seconds, and use them within minutes.

Brand:  Novus Biologicals™ 311-0030

 View more versions of this product

Product Code. 18027913

  • 544.00€ / 1 set
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

For Research Use Only

This item is not returnable. View return policy

Description

Description

  • No desalting or dialysis steps
  • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Rhodamine used in kit has absorption peaks at 520 and 544nm
  • Maximum emission is obtained when the rhodamine is excited at 544nm
Specifications

Specifications

Store at -20°C. Avoid freeze-thaw cycles.
30 μg
Antibody Labeling Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers