Learn More
Novus Biologicals™ Lightning-Link (R) Streptavidin Antibody Labeling Kit
Antibody labeling kit
495.00€ - 975.00€
Specifications
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Streptavidin Antibody Labeling Kit
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18785213
View Documents
Novus Biologicals™
708-0030
30 μg
495.00€
1 set
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
18736483
View Documents
Novus Biologicals™
708-0010
300 μg
774.00€
1 set
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
18745363
View Documents
Novus Biologicals™
708-0015
1 mg
975.00€
1mg
Estimated Shipment: 22-01-2025
Description
Streptavidin is a 53kDa protein purified from Streptomyces avidinii, which has widespread applications due to its very high affinity to the vitamin biotin. Streptavidin is a tetrameric molecule, composed of 4 13kDa monomers, each of which can bind a molecule of biotin. The Lightning-Link Streptavidin kit is unique in the sense that it is the only DIY streptavidin labeling kit available on the market. Like our biotinylation kits, Lightning-Link Streptavidin has been optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step. The way in which the kits are optimised means the antibody or protein of interest will be labeled with up to 3 streptavidin molecules each, maximising the opportunity for biotin to bind and create a biotin streptavidin complex.
Specifications
Specifications
Store at -20°C. Avoid freeze-thaw cycles.
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Streptavidin Antibody Labeling Kit