Learn More
Novus Biologicals™ Lightning-Link (R) Streptavidin Antibody Labeling Kit
Antibody labeling kit
495.00€ - 975.00€
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
Product Type | Streptavidin Antibody Labeling Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785213
|
Novus Biologicals™
708-0030 |
30 μg |
495.00€
1 set |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
18736483
|
Novus Biologicals™
708-0010 |
300 μg |
774.00€
1 set |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
18745363
|
Novus Biologicals™
708-0015 |
1 mg |
975.00€
1mg |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
Streptavidin is a 53kDa protein purified from Streptomyces avidinii, which has widespread applications due to its very high affinity to the vitamin biotin. Streptavidin is a tetrameric molecule, composed of 4 13kDa monomers, each of which can bind a molecule of biotin. The Lightning-Link Streptavidin kit is unique in the sense that it is the only DIY streptavidin labeling kit available on the market. Like our biotinylation kits, Lightning-Link Streptavidin has been optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step. The way in which the kits are optimised means the antibody or protein of interest will be labeled with up to 3 streptavidin molecules each, maximising the opportunity for biotin to bind and create a biotin streptavidin complex.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent | |
Streptavidin Antibody Labeling Kit |