Learn More
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Brand:  Thermo Scientific™ EP0601
 View more versions of this product
Product Code. 10140740
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
Highlights
  • Four minute activation time
  • High PCR specificity and sensitivity
  • Room temperature PCR set up
Product Title
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase > 5U/uL; 100U