Learn More
Deltalab™ Sterile Microtitre Plates
Use Deltalab™ Sterile Microtitre Plates for applications including diluting, serial sample preparation, protein precipitation and hemagglutination. The plates are used when optical reading is required.
Brand:  Deltalab™ 900011
 View more versions of this product
Product Code. 12168910
  • 40.00€ / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Plate dimensions: 127.7 x 85.8 mm.
  • Conical bottom plates may be used for complement fixation
  • Polystyrene lids supplied separately
  • Plates can be covered using sealing mat or the adhesive sealing film
Specifications
Specifications
Standard
“U” form
Flat Bottom (F-well)
50/Pk.
Flat Bottom
Sterile
50