Learn More
Deltalab™ Sterile Microtitre Plates
Use Deltalab™ Sterile Microtitre Plates for applications including diluting, serial sample preparation, protein precipitation and hemagglutination. The plates are used when optical reading is required.
40.00€
Specifications
Sterility Sterile
No. per Pack 50
Quantity 50/Pk.
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12168910
View Documents
Deltalab™
900011
Flat Bottom
40.00€
Pack of 50
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Plate dimensions: 127.7 x 85.8 mm.
  • Conical bottom plates may be used for complement fixation
  • Polystyrene lids supplied separately
  • Plates can be covered using sealing mat or the adhesive sealing film
Specifications
Specifications
Sterile
50
50/Pk.