missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Minisart™ NML Surfactant-free Cellulose Acetate Standard Syringe Filter (SFCA), Sterile
The Minisart™ NML Surfactant-free Cellulose Acetate Standard Syringe Filter (SFCA) is optimal for aqueous solutions with a pH of 4 to 8.
Brand: Sartorius S6555------FMGUK
157.07 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Minisart™ NMFL SFCA syringes are individually packed sterile, making them available for immediate use whenever sterile filtration, clarification and cell removal are needed.
- The (SF)CA membrane is available for the following pore sizes: 0.2 μm and 0.45 μm.
- The housing is made of methacrylate butadiene styrene (MBS), an acrylic resin with limited compatibility with alcohol and free of phthalates.
- Cellulose Acetate Filter
- The cellulose acetate membrane filter combine high flow rates and thermal stability with very low adsorption characteristics are compatible with aqueous solutions, oils, alcohols and other organic solvents.
- Cellulose acetate is also hydrophilic, have due to their symmetric structure high flow rates and is appropriate for aqueous solutions, such as media, buffers and sera.
- Luer Lock Technology
- Having a Luer lock syringe ensures a secure connection, preventing accidental removal of the needle and accidental injection of contents.
- When using a precise laboratory technique, this added quality protection detail is essential for working with complete confidence.
- Wide Variety of Applications
- Due to the low protein binding capacity, low leachables and high flow rate, the Minisart™ NML (SF) CA Syringe Filter has a wide variety of applications.
- It can be used with filtration of aqueous solutions such as cell culture media and serum, sterile filtration and high protein-containing fluids such as plasma.
- Minisart™ Filter Types
- Version K is individually packed and sterilized by ethylene oxide (EO).
- Type S16555 with a pore size of 0.45 μm can be used for clarification and has a male luer lock outlet.
- Type S7594 with a pore size of 5 μm can be used for particle, rubber grit and cell removal and has a male luer lock outlet.
Specifications
Reusable | |
Syringe | |
Cellulose Acetate | |
Sterile | |
Individually Packed | |
1.1 in. | |
Sterile |
CA, Cyrolite | |
MBS | |
0.45 μm | |
50 Pk. | |
Gamma Irridation | |
28 mm |