missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Minisart™ NML Surfactant-free Cellulose Acetate Standard Syringe Filter (SFCA), Sterile
The Minisart™ NML Surfactant-free Cellulose Acetate Standard Syringe Filter (SFCA) is optimal for aqueous solutions with a pH of 4 to 8.
170.00€
Specifications
Disposable | Reusable |
---|---|
For Use With (Equipment) | Syringe |
Material | MBS |
Material (Membrane) | Cellulose Acetate |
Sterility | Sterile |
Product Code | Brand | Pore Size | Sterilization Method | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Sterilization Method | Price | Quantity & Availability | ||||
17103531
|
Sartorius
S6534------FMGUK |
0.2 μm | Gamma Irridation |
170.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
17103541
|
Sartorius
S7597------FXOSK |
0.2 μm | Ethylene Oxide (EO) |
170.00€
Pack of 50 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
17143531
|
Sartorius
S6555------FMOSK |
0.45 μm | Ethylene Oxide (EO) |
170.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
17133531
|
Sartorius
S6555------FMGUK |
0.45 μm | Gamma Irridation |
170.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
17183531
|
Sartorius
S7594------FMOSK |
5 μm | Ethylene Oxide (EO) |
170.00€
Pack of 50 |
|
||||
Description
- Minisart™ NMFL SFCA syringes are individually packed sterile, making them available for immediate use whenever sterile filtration, clarification and cell removal are needed.
- The (SF)CA membrane is available for the following pore sizes: 0.2 μm and 0.45 μm.
- The housing is made of methacrylate butadiene styrene (MBS), an acrylic resin with limited compatibility with alcohol and free of phthalates.
- Cellulose Acetate Filter
- The cellulose acetate membrane filter combine high flow rates and thermal stability with very low adsorption characteristics are compatible with aqueous solutions, oils, alcohols and other organic solvents.
- Cellulose acetate is also hydrophilic, have due to their symmetric structure high flow rates and is appropriate for aqueous solutions, such as media, buffers and sera.
- Luer Lock Technology
- Having a Luer lock syringe ensures a secure connection, preventing accidental removal of the needle and accidental injection of contents.
- When using a precise laboratory technique, this added quality protection detail is essential for working with complete confidence.
- Wide Variety of Applications
- Due to the low protein binding capacity, low leachables and high flow rate, the Minisart™ NML (SF) CA Syringe Filter has a wide variety of applications.
- It can be used with filtration of aqueous solutions such as cell culture media and serum, sterile filtration and high protein-containing fluids such as plasma.
- Minisart™ Filter Types
- Version K is individually packed and sterilized by ethylene oxide (EO).
- Type S16555 with a pore size of 0.45 μm can be used for clarification and has a male luer lock outlet.
- Type S7594 with a pore size of 5 μm can be used for particle, rubber grit and cell removal and has a male luer lock outlet.
Specifications
Reusable | |
MBS | |
Sterile | |
Individually Packed | |
28 mm |
Syringe | |
Cellulose Acetate | |
50 Pk. | |
1.1 in. | |
Sterile |