Learn More
Memmert™ MobileALERT For Up to 4 Alarm Notifications
MobileALERT for 2 alarm notifications, temperature and humidity
Brand:  Memmert™ ICO240C4
 View more versions of this product
Product Code. 17189984
  • 1780.00€ / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • Notification by SMS, temperature and CO2 alarm standard
  • Additionally humidity alarm when equipped with option K7
  • O2 alarm when equipped with option T6
Specifications
Specifications
Temperature and humidity, up to 4 alarms
ICO240
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ MobileALERT For Up to 4 Alarm Notifications > ICO240