Learn More
Memmert™ MobileALERT For Up to 4 Alarm Notifications
MobileALERT for 2 alarm notifications, temperature and humidity
1780.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17110025
|
Memmert™
ICO50C4 |
ICO50 |
1780.00€
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
17119924
|
Memmert™
ICO105C4 |
ICO105 |
1780.00€
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
17149954
|
Memmert™
ICO150C4 |
ICO150 |
1780.00€
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
17189984
|
Memmert™
ICO240C4 |
ICO240 |
1780.00€
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
Description
- Accessory
- Notification by SMS, temperature and CO2 alarm standard
- Additionally humidity alarm when equipped with option K7
- O2 alarm when equipped with option T6
Specifications
Temperature and humidity, up to 4 alarms |
Product Title
Memmert™ MobileALERT For Up to 4 Alarm Notifications