Learn More
Motic™ Moticam A2
Give yourself a head start into digital microscopy with steady 30 fps and a 1080p resolution. The Moticam A2 is a great choice for anyone who wants to begin their journey into live imaging and sharing their discoveries.
Brand: Motic™ 1100600101192
This item is not returnable.
View return policy
Description
- Live Display Modes: 1920 x 1080 px, 1280 x 720 px
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
Specifications
Camera | |
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap | |
1/3.1 in. |
Moticam A | |
sCMOS | |
30 fps |
Product Content Correction
Product Title
Motic™ Moticam A2 > Moticam A2; Frequency (Frame Rate): 30 fps; Sensor Types: sCMOS; Product Line: Moticam A Series; Includes: Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap