missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Aprotinin, Cell Culture Reagent
From bovine lung
Brand: MP Biomedicals™ 0219455983
This item is not returnable.
View return policy
Description
Activity: ∽4 Inhibitor units/mg of dry weight. One Inhibitor unit = 1500 KIU. Lyophilized white powder.
Specifications
Molecular Biology, Radioimmunoassays | |
Aprotinin from Bovine Lung | |
250 mg | |
4°C |
6511.23 | |
5 to 7 | |
Bovine Lung | |
Lyophilized |