missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Aprotinin Protein

From bovine lung

317.00€

Specifications

For Use With (Application) Molecular Biology
Molecular Weight (g/mol) 6511.23
Name Aprotinin from Bovine Lung
pH Range 5 to 7
Source Bovine Lung
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11420662
View Documents
MP Biomedicals™
0219410125
25 mg
317.00€
25mg
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
 
Description

Description

Protease Inhibitor. Activity: ≥3 TIU/mg. Unit Definition: One trypsin inhibitor unit (TIU), which is equal to 900 kallikrein inhibitor units (KIU) and 8800 Schwert and Takenake units (STU), is the amount of inhibitor per mg which hydrolyzes one μmol of benzoyl-DL-arginine-p-nitroanilide (BAPNA) per minute. One TIU reduces the activity of two trypsin units by 50%. It is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, and leukocyte and tissue proteinases.

Spécification

Spécification

Molecular Biology
Aprotinin from Bovine Lung
Bovine Lung
Lyophilized
6511.23
5 to 7
4°C
Vidéos
FDS
Documents

Documents

Certificats