missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lipoprotein-Cholesterol Concentrate

From bovine serum

265.00€ - 731.00€

Specifications

Concentration 1000 to 2000mg/dL
For Use With (Application) Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations
Name Lipoprotein Cholesterol concentrate
pH Range 7.5 to 8
Source Bovine Serum
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11454702
View Documents
MP Biomedicals™
0219147625
25 mL
265.00€
25mL
Please to purchase this item. Need a web account? Register with us today!
11464702
View Documents
MP Biomedicals™
0219147680
100 mL
731.00€
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A mixture of lipoprotein cholesterol, phospholipids and fatty acids. For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or nonenzymatic cholesterol determinations. Offered as a low-salt solution. No preservatives. Free of enzyme inhibitors. Filtered through 0.2ÎĽm membrane filter. Cholesterol concentration: 1000-2000mg/dL.

Specifications

Specifications

1000 to 2000mg/dL
Lipoprotein Cholesterol concentrate
Bovine Serum
Solution
Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations
7.5 to 8
0° to 5°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Lipoprotein-Cholesterol Concentrate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.