Learn More
MP Biomedicals™ Pepstatin A
Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
Brand:  MP Biomedicals™ 0219536810
114.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11495322
  • 146.00€ / 10mg
Estimated Shipment: 19-02-2025
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation
Pepstatin A
0°C
685.91
10 mg
Powder