missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mycoplasma hyopneumoniae Detection Kit

Rapidly detect the antibodies to Mycoplasma hyopneumoniae in porcine serum with this assay.

Brand:  Thermo Scientific™ K004321-9


 View more versions of this product

Product Code. 12325793

  • 4330.00€ / 96 tests
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Detect the antibodies to Mycoplasma hyopneumoniae in porcine serum with Thermo Scientific™ Oxoid™ Mycoplasma hyponeumoniae Detection Kit. The enzyme immunoassay is designed to deliver specific, sensitive, and fast detection of Mycoplasma hyopneumoniae in porcine serum.

Enzootic pneumonia is a disease of pigs, caused by infection with M. hyopneumoniae. A specific and sensitive serological test for the detection of antibodies to M. hyopneumoniae in porcine serum is required for surveillance and precise diagnosis of the disease. The traditional methods for laboratory diagnosis of enzootic pneumonia are complement fixation, indirect haemagglutination, and more recently, indirect ELISA. None of these methods are, however, ideal due to low sensitivity and/or specificity.

Use Mycoplasma hyponeumoniae Detection Kit for the sensitive and specific detection of Mycoplasma hyopneumoniae in porcine samples.


  • Simple: Ready-to-use liquid reagents and concentrates make the assay easy to implement and run, even with high number of samples
  • Sensitive: Detect proteins at low levels for improved assurance of safety
  • Specific: Improved specificity due to selection of highly specific monoclonal antibody against a conserved epitope of the M. hyopneumoniae 74KDa protein.


The Mycoplasma hyopneumoniae ELISA is a blocking assay that has a significantly improved performance compared with traditional assays and is rapid and simple to use even with high numbers of samples.1, 2 The specificity of the test is improved over other methods by the use of a highly specific monoclonal antibody against a conserved epitope of the M. hyopneumoniae 74KDa protein which does not cross-react with M. flocculare.

For Veterinary Use Only.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mycoplasma hyopneumoniae Detection Kit
ELISA (Standard)
960 Tests (10 Plates)
Antibodies to Mycoplasma hyopneumoniae
Antigen coated microwells, Negative control serum, Positive control serum, Sample diluent, Stop solution, Substrate, Wash buffer concentrate
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360D-May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.