Learn More
N-Benzylmaleamic acid, 98+%, Thermo Scientific™
23.30€ - 222.00€
Chemical Identifiers
CAS | 15329-69-8 |
---|---|
Molecular Formula | C11H11NO3 |
Molecular Weight (g/mol) | 205.213 |
MDL Number | MFCD00020482 |
InChI Key | BHWGQIYJCMMSNM-SREVYHEPSA-N |
Synonym | n-benzylmaleamic acid, z-4-benzylamino-4-oxobut-2-enoic acid, 2z-3-benzylcarbamoyl prop-2-enoic acid, 2e-4-benzylamino-4-oxobut-2-enoic acid, n-benzyl maleamic acid, maleic acid monobenzylamide, z-3-benzylcarbamoyl-acrylic acid, 2z-3-n-benzylcarbamoyl prop-2-enoic acid, 2z-4-benzylamino-4-oxobut-2-enoic acid |
PubChem CID | 1549764 |
IUPAC Name | (Z)-4-(benzylamino)-4-oxobut-2-enoic acid |
SMILES | C1=CC=C(C=C1)CNC(=O)C=CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415387
|
Thermo Scientific Alfa Aesar
B22726.06 |
5 g | |||||||
11425387
|
Thermo Scientific Alfa Aesar
B22726.14 |
25 g |
75.10€
25g |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
11435387
|
Thermo Scientific Alfa Aesar
B22726.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
15329-69-8 | |
205.213 | |
BHWGQIYJCMMSNM-SREVYHEPSA-N | |
1549764 | |
C1=CC=C(C=C1)CNC(=O)C=CC(=O)O |
C11H11NO3 | |
MFCD00020482 | |
n-benzylmaleamic acid, z-4-benzylamino-4-oxobut-2-enoic acid, 2z-3-benzylcarbamoyl prop-2-enoic acid, 2e-4-benzylamino-4-oxobut-2-enoic acid, n-benzyl maleamic acid, maleic acid monobenzylamide, z-3-benzylcarbamoyl-acrylic acid, 2z-3-n-benzylcarbamoyl prop-2-enoic acid, 2z-4-benzylamino-4-oxobut-2-enoic acid | |
(Z)-4-(benzylamino)-4-oxobut-2-enoic acid |
Specifications
15329-69-8 | |
C11H11NO3 | |
1212733 | |
BHWGQIYJCMMSNM-SREVYHEPSA-N | |
(Z)-4-(benzylamino)-4-oxobut-2-enoic acid | |
1549764 | |
≥98% |
136°C to 138°C | |
MFCD00020482 | |
n-benzylmaleamic acid, z-4-benzylamino-4-oxobut-2-enoic acid, 2z-3-benzylcarbamoyl prop-2-enoic acid, 2e-4-benzylamino-4-oxobut-2-enoic acid, n-benzyl maleamic acid, maleic acid monobenzylamide, z-3-benzylcarbamoyl-acrylic acid, 2z-3-n-benzylcarbamoyl prop-2-enoic acid, 2z-4-benzylamino-4-oxobut-2-enoic acid | |
C1=CC=C(C=C1)CNC(=O)C=CC(=O)O | |
205.213 | |
205.21 | |
N-Benzylmaleamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 239-361-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures