Learn More
Macherey-Nagel™ N9 Polypropylene Yellow Screw Cap
Brand:  Macherey-Nagel™ 702109
Product Code. 15164722
  • 32.70€ / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Yellow
Silicone white / PTFE blue,
slitted
N9
Polypropylene, PTFE, Silicone
1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ N9 Polypropylene Yellow Screw Cap > Septum: Silicone white / PTFE blue, slitted