missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ APO-DIRECT (TUNEL) Apoptosis Kit

Applications: Flow Cytometry

Brand:  Novus Biologicals™ NBP2-31159

Product Code. 18736133

  • 639.00€ / 1 set
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Positive Control Cells, Negative Control Cells, Wash Buffer, Reaction Buffer, TdT Enzyme, Rinsing Buffer, FITC dUTP, Propidium Iodide/RNase DNA staining solution
APO-DIRECT (TUNEL) Apoptosis Kit
1 Kit
Flow Cytometry
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers