missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ribonuclease Inhibitor Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP1-72417-100UG
This item is not returnable.
View return policy
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-155 of E.coli Ribonuclease Inhibitor The Recombinant E. coli Ribonuclease Inhibitor Protein is derived from E. coli. The Recombinant E. coli Ribonuclease Inhibitor Protein has been validated for the following applications: SDS-Page.
Specifications
SDS-PAGE | |
20kDa | |
100 μg | |
Store at -80°C. Avoid freeze-thaw cycles. | |
Ribonuclease Inhibitor | |
>95% |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT. | |
SDS-PAGE | |
Bacteria | |
RUO | |
Unconjugated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Ribonuclease Inhibitor Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction