missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSnap cfDNA kit for cell-free DNA from plasma

Isolation of cell-free DNA from up to 10 mL blood plasma, utilizing a new column design for high-speed vacuum processing (e.g. with the NucleoVac 24 Vacuum Manifold). Easy-to-use snap-off columns produce ready-to-use, transfection- and molecular-grade DNA with no alcohol precipitation necessary.

205.00€ - 892.00€

Specifications

Format Snap-off Column
Isolation Technology Silica Membrane Technology
Sample Type Plasma, Urine
Elution Volume Elution volume: 20 - 100 μL
Final Product Type Cell-free DNA
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15628455
View Documents
Macherey-Nagel™
740300.10
10
205.00€
10 reactions
Please to purchase this item. Need a web account? Register with us today!
15638455
View Documents
Macherey-Nagel™
740300.50
50
892.00€
50 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The NucleoVac 24 Vacuum Manifold allows the processing of silica columns for nucleic acid purification by vacuum.

  • Snap-off column design for quick processing of large sample volumes
  • Compatible with the NucleoVac 24 vacuum manifold (sold separately, MPN 740299
  • No need for carrier-RNA
  • Process: DNA precipitation—filtration—washing—elution
  • Optimized protocol for use with EDTA or Cell-Free DNA BCT™ (Streck) blood draw tubes
  • Highly efficient recovery of cell-free DNA from Liquid Biopsies
  • No alcohol precipitation necessary
  • Typical downstream applications: NGS, qPCR, ddPCR
Specifications

Specifications

Snap-off Column
Plasma, Urine
Cell-free DNA
DNA isolation from blood plasma
Fragment size: ≥ 50 bp
45 min/6 preps
Silica Membrane Technology
Elution volume: 20 - 100 μL
Vacuum (e.g. NucleoVac 24 Vacuum Manifold and vacuum pump)
NucleoSnap DNA Plasma Columns, Collection Tubes, buffers, Proteinase K
1 - 5 mL
Depends on sample source, storage, and quality
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ NucleoSnap cfDNA kit for cell-free DNA from plasma

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.