missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Tissue Columns

Separate Mini spin columns designed for use with NucleoSpin™ Tissue kits

102.00€ - 420.00€

Specifications

Cell Type Animal & Human tissue, cells
Capacity 60 μg
Column Type Mini spin column
Content And Storage 18°C to 25°C
Description Columns for purification of DNA from a broad range of samples
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15860968
View Documents
Macherey-Nagel™
740952.50S
50 columns
102.00€
Pack of 50
In Stock
to see stock.
Add to basket
 
15850968
View Documents
Macherey-Nagel™
740952.250S
250 columns
420.00€
Pack of 250
In Stock
to see stock.
Add to basket
 
Description

Description

  • NucleoSpin™ Tissue Columns

    • Separate Columns with high DNA recovery and purity
    • Reliable DNA purification for reproducible results
Specifications

Specifications

Animal & Human tissue, cells
Mini spin column
Columns for purification of DNA from a broad range of samples
Mini Spin Column
Animal and Human Tissue, Cells
DNA
Genotyping, Multiplex PCR, Next Generation Sequencing
DNA purification
DNA purification columns
<25 mg human/animal tissue, 102-107 cells
200 bp to 50 kbp
DNA
20 min/prep (excl. lysis)
60 μg
18°C to 25°C
Silica-membrane Technology
Silica Membrane Technology
60 to 100 μL
Centrifugation, Vacuum
NucleoSpin Tissue Columns
DNA
A260/A280 1.7 - 1.9
Room Temperature
Animal & Human tissue, cells
Silica-membrane technology
20 to 35 μg
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 27 months after production

Product Identifier
  • NucleoSpin Tissue

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol

SDS
Documents

Documents

Product Certifications

For research use only