missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Total RNA FFPE XS Column

For rapid isolation of RNA from formalin-fixed, paraffin-embedded samples

83.25€ - 379.00€

Specifications

Format Mini Spin Column
Final Product Type NucleoSpin™ Total RNA FFPE XS Column
For Use With (Application) Rapid isolation of RNA from formalin-fixed, paraffin-embedded samples
Product Type NucleoSpin™ Total RNA FFPE XS Column
Quantity 1 Each
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
11922502
View accessory products View Documents
Macherey-Nagel™
740969.10
10
83.25€
10 reactions
In Stock
to see stock.
Add to basket
 
12768622
View accessory products View Documents
Macherey-Nagel™
740969.50
50
379.00€
50 reactions
In Stock
to see stock.
Add to basket
 
Description

Description

NucleoSpin FFPE RNA, no restriction for total RNA isolation - even from FFPE samples.
Enhance your RT-PCR* performance. Improved template quality - get less fragmented, well decrosslinked, DNA-free RNA.
Decrosslinking buffer and rDNase provided with the kit. Improved sensitivity - highly concentrated RNA by elution in only 5μL to 30μL. Use of the XS (mini elution volume) column design.
Convenient paraffin removal
Simply melt paraffin with the Paraffin Dissolver (patent pending) provided with the kit. Without toxic organic solvent - without time-consuming mixing steps - without long incubation time.
Save time
Minimum number of working steps. Fast procedure preserves RNA quality.
Suitable for a large variety of starting materials, like clinical samples
e.g., colon carcinoma (hematoxylin stained/unstained), colon mucosa, adenocarcinoma of colon transversum, liver (rat/human), liver carcinoma (hematoxylin stained), lymph node, spleen (rat/human); pancreas (rat/human; diabetic/nondiabetic), placenta.
  • Silica membrane technology
  • Format: mini spin columns - XS design for increased sensitivity
  • Sample material: up to 7 sections of 250mm², or 15mg
  • RNA yield depends on the amount and quality of the sample
  • Typical ration A260/A280: 1.9 to 2. 1
  • Typical RIN: 2 to 6
  • Typical RNA size: 100nt to 5, 00nt
  • Elution volume: 5μL to 30μL
  • Preparation time: 70min
  • Easy paraffin removal
  • Convenient handling
  • Fast procedure RNA purification from FFPE samples.
Specifications

Specifications

Mini Spin Column
Rapid isolation of RNA from formalin-fixed, paraffin-embedded samples
1 Each
NucleoSpin™ Total RNA FFPE XS Column
NucleoSpin™ Total RNA FFPE XS Column
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 1,4-dioxane, proteinase K, paraffin dissolver, rDNase

SDS
Documents

Documents

Product Certifications