missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NUDT2 (Human) Recombinant Protein
Human NUDT2 full-length ORF ( AAH04926, 1 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000318-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MALRACGLIIFRRCLIPKVDNNAIEFLLLQASDGIHHWTPPKGHVEPGEDDLETALRETQEEAGIEAGQLTIIEGFKRELNYVARNKPKTVIYWLAEVKDYDVEIRLSHEHQAYRWLGLEEACQLAQFKEMKAALQEGHQFLCSIEA
Specifications
AAH04926 | |
nudix (nucleoside diphosphate linked moiety X)-type motif 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NUDT2 | |
GST |
318 | |
Wheat germ expression system | |
25 μg | |
APAH1, MGC10404 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |