Learn More
Usbeck Laborgeraete™ Nylon Universal Scissors
Brand:  Usbeck Laborgeraete™ 4280
 View more versions of this product
Product Code. 10565292
  • 11.10€ / Each
Estimated Shipment: 12-05-2025
Specifications
Universal Scissors
170 mm
Nylon
Product Title
Usbeck Laborgeraete™ Nylon Universal Scissors > Length: 170mm