missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ OIP5 Recombinant Protein

Brand:  Abnova™ H00011339-P01.10ug

 View more versions of this product

Product Code. 16196572

  • 335.00€ / 10µg
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH15050
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
51.3
8
Glutathione Sepharose 4 Fast Flow
10 μg
MAAQPLRHRSRCATPPRGDFCGGTERAIDQASFTTSMEWDTQVVKGSSPLGPAGLGAEEPAAGPQLPSWLQPERCAVFQCAQCHAVLADSVHLAWDLSRSLGAVVFSRVTNNVVLEAPFLVGIEGSLKGSTYNLLFCGSCGIPVGFHLYSTHAALAALRGHFCLSSDKMVCYLLKTKAIVNASEMDIQNVPLSEKIAELKEKIVLTHNRLKSLMKILSEVTPDQSKPEN
5730547N13Rik/LINT-25/MIS18beta
OIP5
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
11339
OIP5 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OIP5
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers