Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Brand: Thermo Scientific™ D2-GK
This item is not returnable.
View return policy
Description
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
Specifications
Replacement Gaskets (1 pair) | |
Owl™ D2 Wide Gel Electrophoresis System | |
Owl | |
Gasket |