Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Brand:  Thermo Scientific™ D2-GK
 View more versions of this product
Product Code. 12424607
  • 25.55€ / Each
Estimated Shipment: 13-03-2025
Description
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Gaskets (1 pair)
Owl™ D2 Wide Gel Electrophoresis System
Owl
Gasket