Learn More
DiscoverX PathHunter™ CHO-K1 DRD2S (Short Isoform) β-Arrestin Cell Line
The PathHunter™ CHO-K1 DRD2S (Short Isoform) β-Arrestin Cell Line measures DRD2S (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0351C2
Encompass
Product Code. 17393019
  • 33718.50€ / 2 ampoules
Estimated Shipment: 17-01-2025
Description
The PathHunter™ DRD2S β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD2S GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged DRD2S and EA-tagged β-Arrestin 2. Activation of DRD2S stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
β-Arrestin recruitment
Cell Lines
2 Vials
NM_016574.3
CHO-K1
Dopamine HCl
DRD2S
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.