Learn More
DiscoverX PathHunter™ CHO-K1 F2RL3 (PAR4) β-Arrestin Cell Line
The PathHunter™ CHO-K1 F2RL3 (PAR4) β-Arrestin Cell Line measures F2RL3 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0279C2
Encompass
Product Code. 17382889
  • 35218.50€ / 2 ampoules
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The PathHunter™ F2RL3 (PAR4) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serine proteases) based activation of F2RL3 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged F2RL3 and EA-tagged β-Arrestin 2. Activation of F2RL3 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
CHO-K1
Protease-Activated Receptor-4 (AYPGKF-NH2)
F2RL3 (PAR4)
Cell Lines
2 Vials
AY431102