Learn More
DiscoverX PathHunter™ CHO-K1 mCCR3 β-Arrestin Cell Line
The PathHunter™ CHO-K1 mCCR3 β-Arrestin Cell Line measures mCCR3 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Mouse protein.
Brand:  DiscoverX 93-0522C2
Encompass
Product Code. 17393319
  • 35218.50€ / 2 ampoules
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The PathHunter™ mCCR3 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL24) based activation of mCCR3 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mCCR3 and EA-tagged β-Arrestin 2. Activation of mCCR3 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.
Specifications
Specifications
β-Arrestin recruitment
Cell Lines
2 Vials
NM_009914
CHO-K1
Recombinant Murine Eotaxin-2 (CCL24)
mCCR3