Learn More
DiscoverX PathHunter™ CHO-K1 mHCRTR1 β-Arrestin Cell Pool
The PathHunter™ CHO-K1 mHCRTR1 β-Arrestin Cell Pool measures mHCRTR1 (GPCR) activity via recruitment of β-Arrestin2. This cell pool ships as two vials of cryopreserved cells. Mouse protein.
Brand:  DiscoverX 93-0606P2
Encompass
Product Code. 17363519
  • 14968.50€ / 2 ampoules
Estimated Shipment: 17-01-2025
Description
The PathHunter™ mHCRTR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. orexin A) based activation of mHCRTR1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mHCRTR1 and EA-tagged β-Arrestin 2. Activation of mHCRTR1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the mouse version of this protein.
Specifications
Specifications
β-Arrestin recruitment
Cell Pools
mHCRTR1
CHO-K1
2 Vials
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.