Learn More
DiscoverX PathHunter™ CHO-K1 mOPRD1 β-Arrestin Cell Line
The PathHunter™ CHO-K1 mOPRD1 β-Arrestin Cell Line measures mOPRD1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Mouse protein.
Brand: DiscoverX 93-0611C2
This item is not returnable.
View return policy
Description
The PathHunter™ mOPRD1 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. DADLE) based activation of mOPRD1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mOPRD1 and EA-tagged β-Arrestin 2. Activation of mOPRD1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.Specifications
β-Arrestin recruitment | |
Cell Lines | |
2 Vials | |
NM_013622.3 |
CHO-K1 | |
[D-Ala2, D-Leu5]-Enkephalin (DADLE) | |
mOPRD1 |