Learn More
DiscoverX PathHunter™ CHO-K1 mOPRD1 β-Arrestin Cell Line
The PathHunter™ CHO-K1 mOPRD1 β-Arrestin Cell Line measures mOPRD1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Mouse protein.
Brand:  DiscoverX 93-0611C2
Encompass
Product Code. 17303529
  • 35218.50€ / 2 ampoules
Estimated Shipment: 03-02-2025
Description
The PathHunter™ mOPRD1 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. DADLE) based activation of mOPRD1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mOPRD1 and EA-tagged β-Arrestin 2. Activation of mOPRD1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.
Specifications
Specifications
β-Arrestin recruitment
Cell Lines
2 Vials
NM_013622.3
CHO-K1
[D-Ala2, D-Leu5]-Enkephalin (DADLE)
mOPRD1
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.